Learn More
MilliporeSigma™ Supelco™ Magnetic Screw Cap for Headspace Vials
Catalog No. 111020921 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color:
Gray, Red
White, Blue
White, Blue Transparent
Septum:
PTFE-faced Butyl
PTFE/Silicone
Thickness (Metric):
1.3 mm
1.5 mm
1.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Thickness (Metric)
11-102-0921 Gray, Red PTFE-faced Butyl 1.6 mm
11-102-0858 White, Blue PTFE/Silicone 1.5 mm
11-101-0783 White, Blue Transparent PTFE/Silicone 1.3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-0921 Supplier MilliporeSigma™ Supelco™ Supplier No. SU860102
Add to Cart
Add to Cart
This magnetic screw cap has been developed for the CTC instrument.
  • The vial has a precision thread with numerous threads to force the septa against the vial to seal it to be gas tight.
  • The center hole is designed to be used for SPME as well as standard Headspace analysis, but still offers enough surface for the magnet to transport a completely filled vial.
TRUSTED_SUSTAINABILITY
Specifications
Color Gray, Red
Septum PTFE-faced Butyl
No. per Pack 100
Thickness (Metric) 1.6 mm
Closure Size 18 mm