Learn More
V&P Scientific Inc MAGNETIC RACK, 1 Flip Top Microcentrifuge Tube or Conical Tube, 5mL or 15mL Volume
Catalog No. NC1787836
Encompass
MAGNETIC BEAD SEPARATION RACK, 1 Flip Top Microcentrifuge Tube or Conical Tube, 5mL or 15mL Volume, 1 Sided Bead Pellet Location, Low Position, 42MGO, NdFeB Magnet, 17mm Hole Diameter, Resin
Catalog No. NC1787836 Supplier V&P Scientific Inc Supplier No. VP 772F32
Add to Cart
Add to Cart
MAGNETIC BEAD SEPARATION RACK, 1 Flip Top Microcentrifuge Tube or Conical Tube, 5mL or 15mL Volume, 1 Sided Bead Pellet Location, Low Position, 42MGO, NdFeB Magnet, 17mm Hole Diameter, Resin