Learn More
IBA LifeSciences Magnetic Separator for magnetic beads from solution
Catalog No. NC1540242
Encompass
The Magnetic Separator is a transparent plastic rack with 4 x 12 wells – two rows for 2 ml reaction tubes (left side) and 1.5 ml reaction tubes (right side), respectively. The middle rows contain magnets that are suitable for the separation of Strep-TactinXT-coated magnetic beads, MagStrep "type3" XT beads, within seconds from a solution making the Magnetic Separator a helpful tool for efficient, easy, and fast batch purification of Strep-tagII and Twin-Strep-tag fusion proteins. The separation procedure can be interrupted by placing the reaction tube in one of the correspondent outer rows.
Catalog No. NC1540242 Supplier IBA LifeSciences Supplier No. 21602000
The Magnetic Separator is a transparent plastic rack with 4 x 12 wells – two rows for 2 ml reaction tubes (left side) and 1.5 ml reaction tubes (right side), respectively. The middle rows contain magnets that are suitable for the separation of Strep-TactinXT-coated magnetic beads, MagStrep "type3" XT beads, within seconds from a solution making the Magnetic Separator a helpful tool for efficient, easy, and fast batch purification of Strep-tagII and Twin-Strep-tag fusion proteins. The separation procedure can be interrupted by placing the reaction tube in one of the correspondent outer rows.