Learn More
Electron Microscopy Sciences Magnetic Bead Separation Rack for Std Flat Microplate
SDP
Catalog No. 501928540
Encompass_Preferred
Magnetic bead separation racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays. They offer exceptional quality and value compared to competing brands. Racks fit microplates following ANSI standards. Clear acrylic allows unobstructed view of tube contents. Magnets covering entire footprint to accept all standard micro-plates.
Catalog No. 50-192-8540 Supplier Electron Microscopy Sciences Supplier No. 6860555
Magnetic bead separation racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays. They offer exceptional quality and value compared to competing brands. Racks fit microplates following ANSI standards. Clear acrylic allows unobstructed view of tube contents. Magnets covering entire footprint to accept all standard micro-plates.