Learn More
World Precision Instrument M1 Magnetic Stand
SDP
Catalog No. 50822961
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
A precision magnetic base provides stable support for such devices as electrodes and micromanipulators. The adjustable second arm adopts a variety of angles.These are an ideal manipulator stands.
Optional M5 10mm clamp should be added for use with M3301, DC3001, MD4 and MMJ manipulators.
Catalog No. 50-822-961 Supplier World Precision Instrument Supplier No. M1
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
A precision magnetic base provides stable support for such devices as electrodes and micromanipulators. The adjustable second arm adopts a variety of angles.These are an ideal manipulator stands.
Optional M5 10mm clamp should be added for use with M3301, DC3001, MD4 and MMJ manipulators.