Learn More
World Precision Instrument Magnetic Stand Rotatable
SDP
Catalog No. 50822974
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
Mechanical clamp tightens three rotatable joints simultaneously with one locking knob. Arm adjusts without distortion. Base exerts a magnetic force of 100 kg for greatest stability. Equipped with fine adjustment for precise operations.
Catalog No. 50-822-974 Supplier World Precision Instrument Supplier No. M9
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
Mechanical clamp tightens three rotatable joints simultaneously with one locking knob. Arm adjusts without distortion. Base exerts a magnetic force of 100 kg for greatest stability. Equipped with fine adjustment for precise operations.