Learn More
World Precision Instrument Magnetic Stand with Balljoint
SDP
Catalog No. 50822973
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
A ball joint at the base of the main post allows 360° rotation, offering considerable versatility. The second arm adopts angles up to 75°. (Optional M5 10 mm clamp should be added for use with M3301, DC3001, MD4 and MMJ manipulators.)
Catalog No. 50-822-973 Supplier World Precision Instrument Supplier No. M8
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
A ball joint at the base of the main post allows 360° rotation, offering considerable versatility. The second arm adopts angles up to 75°. (Optional M5 10 mm clamp should be added for use with M3301, DC3001, MD4 and MMJ manipulators.)