Learn More
World Precision Instrument M10L Magnetic Stand
SDP
Catalog No. 50822963
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
This magnetic base is the same as the M10 (12mm diameter sub pole), but it is equipped with a taller (14-inch) vertical main pole. These are an ideal manipulator stands.
Catalog No. 50-822-963 Supplier World Precision Instrument Supplier No. M10L
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
This magnetic base is the same as the M10 (12mm diameter sub pole), but it is equipped with a taller (14-inch) vertical main pole. These are an ideal manipulator stands.