Learn More
World Precision Instrument Magnetic Stand - Flexible
SDP
Catalog No. 50822964
Encompass
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
The arm of this magnetic base bends freely for maximum flexibility. The connecting arm twists and bends like a snake. Lock the arm in position with a flick of the controlling lever. These are an ideal stands probe holders and lightweight objects.
Catalog No. 50-822-964 Supplier World Precision Instrument Supplier No. M11
The base of each stand exerts a powerful magnetic force that holds it solidly on ferrous metal surfaces - even vertically or upside down
The arm of this magnetic base bends freely for maximum flexibility. The connecting arm twists and bends like a snake. Lock the arm in position with a flick of the controlling lever. These are an ideal stands probe holders and lightweight objects.