Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Magnetic Stand for 12 Tubes (1.5mL, 2mL)

Catalog No. 502719181
Encompass_Preferred

Magnetic Stand for 12 Tubes (1.5mL, 2mL)

Catalog No. 50-271-9181 Supplier ACROBiosystems Supplier No. MB021EA
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnetic Stand for 12 Tubes (1.5mL, 2mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.