Learn More
MilliporeSigma™ Supelco™ Magnetic Stir Bar
Catalog No. 111021611 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
0.1 to 2 mL Vessels
3 to 5 mL Vessels
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-102-1611 3 to 5 mL Vessels
11-102-1610 0.1 to 2 mL Vessels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1611 Supplier MilliporeSigma™ Supelco™ Supplier No. 23227
Add to Cart
Edge
Add to Cart
Magnetic stir bar may be used for stirring samples in the stir-bar sorptive extraction (SBSE) of volatile components of malt whisky, analyzed using gas chromatography coupled with mass spectrometry (GC-MS)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 3 to 5 mL Vessels
Material PTFE
Quantity 6/Pk.