Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, NdFeB Magnet, 52MGO, PVDF Encapsulated, Used in Bottles, 10/pack
Catalog No. NC1916686
Encompass
STIR ELEMENT, CYLINDER, NdFeB Magnet, 52MGO, PVDF Encapsulated, 11.5mm Diameter, 48.2mm Length, 80°C Max Operating Temp for Magnets
Catalog No. NC1916686 Supplier V&P Scientific Inc Supplier No. VP 772BPN521148
STIR ELEMENT, CYLINDER, NdFeB Magnet, 52MGO, PVDF Encapsulated, 11.5mm Diameter, 48.2mm Length, 80°C Max Operating Temp for Magnets