Learn More
Chemglass Life Sciences Chemglass Plain PTFE Magnetic Stir Bar, 80 x 10 mm
SDP
Catalog No. NC3242821
Encompass_Preferred
Plain PTFE magnetic stir bar for use with laboratory magnetic stir plates. The smooth cylindrical profile provides consistent mixing and helps reduce vortexing across a range of vessel sizes. The PTFE coating offers chemical resistance and easy cleaning, while the encapsulated magnetic core ensures reliable coupling to stir plates.
  • Made from PTFE for broad chemical resistance.
  • Plain cylindrical profile for consistent mixing.
  • Encapsulated magnetic core for reliable coupling to stir plates.
  • Dimensions: 80 x 10 mm (length x diameter).
  • Sold individually (each).
  • Suitable for routine laboratory stirring and agitation.
Catalog No. NC3242821 Supplier Chemglass Life Sciences Supplier No. CG2001P10
Plain PTFE magnetic stir bar for use with laboratory magnetic stir plates. The smooth cylindrical profile provides consistent mixing and helps reduce vortexing across a range of vessel sizes. The PTFE coating offers chemical resistance and easy cleaning, while the encapsulated magnetic core ensures reliable coupling to stir plates.
  • Made from PTFE for broad chemical resistance.
  • Plain cylindrical profile for consistent mixing.
  • Encapsulated magnetic core for reliable coupling to stir plates.
  • Dimensions: 80 x 10 mm (length x diameter).
  • Sold individually (each).
  • Suitable for routine laboratory stirring and agitation.