Learn More
Cole-Parmer Bel-Art Stir Bar Retriever, Flexible PTFE; 14-3/4" Long
Supplier:  Cole-Parmer 0855520
Encompass
Easily retrieve stir bars without spills or possible breakage. The alnico V magnet is completely enclosed in PTFE.
Catalog No. NC9342725