Learn More
Azzota Corp 4 Points Stand-Alone Magnetic Stirrer, 1000Ml Volume Per Point, Four Controller, Speed Range 0-1200Rpm, Materials Stainless Steel, Dimension 150 X 150 X 25 mm
Catalog No. 507532949
Encompass_Preferred
4 Points stand-alone Magnetic stirrer, 1000ml Volume per point, Four controller, Speed range 0-1200RPM, Materials Stainless steel, Dimension 150 x 150 x 25 mm
Catalog No. 50-753-2949 Supplier Azzota Corp Supplier No. MS4X4
4 Points stand-alone Magnetic stirrer, 1000ml Volume per point, Four controller, Speed range 0-1200RPM, Materials Stainless steel, Dimension 150 x 150 x 25 mm