Learn More
Azzota Corp 4 Points Synchronized Magnetic Stirrer, 1000Ml Volume Per Point, One Controller, Speed Range 0-1200Rpm, Materials Stainless Steel, Dimension 150 X 150 X 25 mm
Catalog No. 507532947
Encompass_Preferred
4 Points synchronized Magnetic stirrer, 1000ml Volume per point, One controller, Speed range 0-1200RPM, Materials Stainless steel, Dimension 150 x 150 x 25 mm
Catalog No. 50-753-2947 Supplier Azzota Corp Supplier No. MS4X1
4 Points synchronized Magnetic stirrer, 1000ml Volume per point, One controller, Speed range 0-1200RPM, Materials Stainless steel, Dimension 150 x 150 x 25 mm