Learn More
Azzota Corp Single Point Magnetic Stirrer, 1000Ml Volume Per Point, Speed Range 0-1200Rpm, Materials Stainless Steel, Dimension 150 X 150 X 25 mm
Catalog No. 507532943
Encompass_Preferred
Single Point Magnetic stirrer, 1000ml Volume per point, Speed range 0-1200RPM, Materials Stainless steel, Dimension 150 x 150 x 25 mm
Catalog No. 50-753-2943 Supplier Azzota Corp Supplier No. MS1X1
Single Point Magnetic stirrer, 1000ml Volume per point, Speed range 0-1200RPM, Materials Stainless steel, Dimension 150 x 150 x 25 mm