Learn More
SI Analytics™ Magnetic Stirrer
Catalog No. 15177887 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
For Use With (Equipment):
TitroLine 7000, 7500 KF Trace, 7750, 7800 and TITRONIC 500
Catalog No. For Use With (Equipment)
15-177-887 TitroLine 7000, 7500 KF Trace, 7750, 7800 and TITRONIC 500
1 options
Catalog No. 15-177-887 Supplier SI Analytics™ Supplier No. 285220400
Add to Cart
Add to Cart
For use with YSI TitroLine™ Autotitrators
  • Agitator speed is infinitely adjustable from 500 to 2000 r/min
TRUSTED_SUSTAINABILITY
Specifications
Connector Types TitroLine™ 6000/7000 and TITRONIC™ 500
Container Size Vessels up to 500 mL
Type Magnetic Stirrer
Includes TZ 1855 Power Supply
Model TM 235
Range Speed: 500 to 2000 r/min
For Use With (Equipment) TitroLine 7000, 7500 KF Trace, 7750, 7800 and TITRONIC 500