Learn More
Mettler Toledo™ Magnetic Stirrer EasyMix
Catalog No. 01911932 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyMix
Catalog No. For Use With (Equipment)
01-911-932 EasyMix
1 options
This item is not returnable. View return policy
Catalog No. 01-911-932 Supplier Mettler Toledo™ Supplier No. 30671570
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Magnetic stirrer EasyMix for use with SevenDirect meters. The EasyMix can be controlled manually or automatically by the SevenDirect meter.
  • The Right Stirrer
  • A dedicated stirrer for the SevenDirect generates key advantages: programmable stirring and a perfect fit with the EasyPlace™ electrode arm
  • Program Consistent Conditions
  • Program the stirring speed from the pH meter to ensure consistent conditions for all operators
  • Define preconditioning and waiting periods
  • Perfect Fit Saves Space
  • The EasyMix magnetic stirrer is a perfect fit with the EasyPlace electrode arm
  • Together, they form a tidy and compact setup
  • Reliably Stir Large Volumes
  • The larger the sample, the higher the risk of inhomogeneity
  • Rely on the strength of the EasyMix to stir efficiently up to 10 L of sample volume
TRUSTED_SUSTAINABILITY
Specifications
Type Magnetic Stirrer, EasyMix
For Use With (Equipment) EasyMix