Learn More
Perkin Elmer US LLC Magnetic Stirrer Fleas - Qty. 6
Catalog No. 502112846
Encompass_Preferred
The Magnetic Stirrer Fleas, also called stirring beads, are required with the Quartz SUPRASIL Fluorescence Cells for Magnetic Stirrers (B0631132), but they need to be order separately. With this part number, they are also available as a replacement or spare set. Qty. 6 beads per package.
Catalog No. 50-211-2846 Supplier Perkin Elmer US LLC Supplier No. 04978499
The Magnetic Stirrer Fleas, also called stirring beads, are required with the Quartz SUPRASIL Fluorescence Cells for Magnetic Stirrers (B0631132), but they need to be order separately. With this part number, they are also available as a replacement or spare set. Qty. 6 beads per package.