Learn More
Alkali Scientific Alkali Scientific™ Magnetic Stirrer IPS7101-150 with 150L Mixing Capacity With Chemically Resistant Ceramic Top And Digital Controls, 115V
SDP
Catalog No. 503029520
Encompass_Preferred
Alkali Industrial HCS Series of high-capacity stirrers provide reliable performance and safe operation for high throughput lab use. These stirrers feature a durable, chemically resistant white ceramic top plate, which protects the internals of the stirrer from leaks and spills. The powerful motor drive system ensures efficient and thorough mixing in large vessels and viscous samples. Ideal for large-scale stirring applications, the HCS Series offers 50-liter and 150-liter maximum capacity models, as HCS-50L and HCS-150L respectively. Mixing speed can be viewed on the large digital display, and speed is adjustable from 50 rpm to 1500 rpm (HCS-50L) or 1200 rpm (HCS-150L). The built-in timer settings range from 1 second to 9 hours and include a continuous operation setting. Stir up to 150-liter volumes Chemically resistant white ceramic top surface Powerful motor ideal for large vessel applications Digital control of both stirring speed and timer
Catalog No. 50-302-9520 Supplier Alkali Scientific Supplier No. IPS7101150
Alkali Industrial HCS Series of high-capacity stirrers provide reliable performance and safe operation for high throughput lab use. These stirrers feature a durable, chemically resistant white ceramic top plate, which protects the internals of the stirrer from leaks and spills. The powerful motor drive system ensures efficient and thorough mixing in large vessels and viscous samples. Ideal for large-scale stirring applications, the HCS Series offers 50-liter and 150-liter maximum capacity models, as HCS-50L and HCS-150L respectively. Mixing speed can be viewed on the large digital display, and speed is adjustable from 50 rpm to 1500 rpm (HCS-50L) or 1200 rpm (HCS-150L). The built-in timer settings range from 1 second to 9 hours and include a continuous operation setting. Stir up to 150-liter volumes Chemically resistant white ceramic top surface Powerful motor ideal for large vessel applications Digital control of both stirring speed and timer