Learn More
Thomas Scientific MiniMag Mini Magnetic Stirrer, 100-240V US Plug
SDP
Catalog No. 501458100
Encompass_Preferred
With a square footprint of under 5 inches, the MiniMag™ is the most compact stirrer in the Benchmark line. Despite its modest dimensions, the 3.4 inch square work surface accepts a wide variety of beakers, flasks, bottles and other laboratory vessels for stirring volumes as large as 1.5 liters. The stirring speed is continuously adjustable from 100 to 1500rpm. For temperature sensitive samples, the MiniMag is incubator and cold-room safe for use from +4°C to +45°C. "Mini" sized, less than 3.5" wide Adjustable speed control, 100 to 1500rpm Effectively mixes volumes up to 1.5L
Catalog No. 50-145-8100 Supplier Thomas Scientific Supplier No. 1177X38
With a square footprint of under 5 inches, the MiniMag™ is the most compact stirrer in the Benchmark line. Despite its modest dimensions, the 3.4 inch square work surface accepts a wide variety of beakers, flasks, bottles and other laboratory vessels for stirring volumes as large as 1.5 liters. The stirring speed is continuously adjustable from 100 to 1500rpm. For temperature sensitive samples, the MiniMag is incubator and cold-room safe for use from +4°C to +45°C. "Mini" sized, less than 3.5" wide Adjustable speed control, 100 to 1500rpm Effectively mixes volumes up to 1.5L