Learn More
Electron Microscopy Sciences MagLab MagPette Magnetic Lab Storage, Purple
SDP
Catalog No. 501929497
Encompass_Preferred
With our new MagLab storage system, easily organize common lab products off your bench top. The racks are made of ABS plastic in bright translucent colors. Each piece features four powerful magnets for a secure hold onto most metal surfaces. Or use the metal plate provided to create your own holding point. Holds two pipettors (fits most popular styles).
Color: Purple
Length: 5.1 in
Width: 4.4in
Catalog No. 50-192-9497 Supplier Electron Microscopy Sciences Supplier No. 6438004
With our new MagLab storage system, easily organize common lab products off your bench top. The racks are made of ABS plastic in bright translucent colors. Each piece features four powerful magnets for a secure hold onto most metal surfaces. Or use the metal plate provided to create your own holding point. Holds two pipettors (fits most popular styles).
Color: Purple
Length: 5.1 in
Width: 4.4in