Learn More
OHAUS Magnetic Strip Blot Box
Catalog No. 01804356 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-356 Shaker Attachment
1 options
Catalog No. 01-804-356 Supplier OHAUS Supplier No. 83041371
Add to Cart
Add to Cart
Used in rocking mode.
  • Holds six Western Blot Strips.
  • Attaches to the top or bottom of magnetic platform.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment