Learn More
Mitegen Magnetic Wand with Rubber Grips, For Secure Handling of Goniometer Bases & Loops in Cryocrystallography
SDP
Catalog No. NC1645193
Encompass_Preferred
This Magnetic Wand with Rubber Grips is an essential tool designed to significantly improve the handling of goniometer bases (caps) and loops during demanding cryocrystallography sessions. Engineered for enhanced control and comfort, the addition of secure rubber grips ensures a firm, non-slip hold, minimizing the risk of accidental drops or misplacements of delicate samples and components. This improved design is particularly beneficial during long crystal harvesting sessions, reducing hand fatigue and enhancing precision. The magnetic tip securely engages with compatible goniometer bases and loops, streamlining workflows in cryogenic environments. By providing superior dexterity and stability, this magnetic wand becomes an indispensable aid for researchers and technicians engaged in crystallography, ensuring safer and more efficient sample manipulation from start to finish.
Catalog No. NC1645193 Supplier Mitegen Supplier No. MW1C
This Magnetic Wand with Rubber Grips is an essential tool designed to significantly improve the handling of goniometer bases (caps) and loops during demanding cryocrystallography sessions. Engineered for enhanced control and comfort, the addition of secure rubber grips ensures a firm, non-slip hold, minimizing the risk of accidental drops or misplacements of delicate samples and components. This improved design is particularly beneficial during long crystal harvesting sessions, reducing hand fatigue and enhancing precision. The magnetic tip securely engages with compatible goniometer bases and loops, streamlining workflows in cryogenic environments. By providing superior dexterity and stability, this magnetic wand becomes an indispensable aid for researchers and technicians engaged in crystallography, ensuring safer and more efficient sample manipulation from start to finish.