Learn More
Description
- North and South poles are located on opposite faces for ease of use
- Can be used to determine if an object is magnetic
- A pair of magnetic wands can be used to demonstrate attraction and repulsion between poles
Specifications
Specifications
| Shape | Rectangular |
| Grade | 1+ |
| Color | Blue, Green, Orange, Pink, Red, Violet, Yellow |
| Product Type | Magnet Wand Kit |
| Length (English) | 7.5 in. |
| Quantity | 7 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
