Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnetic Wands
SCI_ED

Catalog No. s98480
Change view
Click to view available options
Grade:
K to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98480 K to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S98480 Supplier Learning Resources, Inc Supplier No. LER0762
Only null left
Add to Cart
Add to Cart

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Introduce children to the magic of magnetism.

  • Set of six wands in a rainbow of colors

Order Info

Grades K to 5

Specifications

For Use With (Application) Introduce children to the magic of magnetism
Grade Level Elementary
Grade K to 5
Color Rainbow
Product Type Magnetic Wand
Age 5 to 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.