Learn More
ATCC Magnetospirillum magneticum
Catalog No. 502381937
Encompass
Magnetospirillum magneticum strain AMB-1 is a bacterial strain isolated from a water pond in Japan. When this strain is grown under microaerophilic conditions, the cells are magnetotactic. They have magnetite inclusions, which cause the cells to turn in a magnetic field.
Catalog No. 50-238-1937 Supplier ATCC Supplier No. 700264
Magnetospirillum magneticum strain AMB-1 is a bacterial strain isolated from a water pond in Japan. When this strain is grown under microaerophilic conditions, the cells are magnetotactic. They have magnetite inclusions, which cause the cells to turn in a magnetic field.