Learn More
Bioworld MagneZoom™ A Protein-A Magnetic Beads, 1 mL
Catalog No. 50197577
Encompass
Recombinant protein-A covalently linked to MagneZoom (magnetic beads). For magnetic separation and isolation of IgG subclasses from ascites, serum, or cell culture supernatants.Technical Specifications:Particle size: 2-5µmLigand: rProtein A Binding Capacity: 0.20mg rabbit IgG/ml beadspH stability: 4-9Supplied as 25mg/ml suspensionRequires MagneZoom paramagnetic bead station (20160000-1) for sepaating beads.Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.
Catalog No. 50-197-577 Supplier Bioworld Supplier No. 201620001
Recombinant protein-A covalently linked to MagneZoom (magnetic beads). For magnetic separation and isolation of IgG subclasses from ascites, serum, or cell culture supernatants.Technical Specifications:Particle size: 2-5µmLigand: rProtein A Binding Capacity: 0.20mg rabbit IgG/ml beadspH stability: 4-9Supplied as 25mg/ml suspensionRequires MagneZoom paramagnetic bead station (20160000-1) for sepaating beads.Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.