Learn More
Bioworld MagneZoom™ CAM (Calmodulin), 2 mL
Catalog No. 50197580
Encompass
Calmodulin covalently bound to MagneZoom magnetic beads. For purification of calmodulin-binding proteins and calmodulin-regulated proteins. Proteins containing a calmodulin binding peptide fusion tag can also be purified using MagneZoom - CAM.Technical SpecificationsParticle Size: 2-5 µmLigand: Bovine CalmodulinSupplied as a suspension, 25 mg/mLpH stability: 4-9Requires MagneZoom™ paramagnetic bead station (20160000-1).Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.
Catalog No. 50-197-580 Supplier Bioworld Supplier No. 201620022
Calmodulin covalently bound to MagneZoom magnetic beads. For purification of calmodulin-binding proteins and calmodulin-regulated proteins. Proteins containing a calmodulin binding peptide fusion tag can also be purified using MagneZoom - CAM.Technical SpecificationsParticle Size: 2-5 µmLigand: Bovine CalmodulinSupplied as a suspension, 25 mg/mLpH stability: 4-9Requires MagneZoom™ paramagnetic bead station (20160000-1).Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.