Learn More
Bioworld MagneZoom™ Control Paramagnetic Beads, 2 mL
Catalog No. 50197597
Encompass
To remove non-specific binding of proteins to magnetic bead surface and to achieve clean co-immunoprecipitation of your desired antibody or any other ligand binding. Use it with any magnetic beads experiment.Size: 2-5µm.Silica-coated magnetic beadspH stability: 4-9Supplied as 25mg/ml suspension.Requires MagneZoom paramagnetic bead station (20160000-1) for separating beads.Usage: bioWorld's products are for LABORATORY RESEARCH use only.
Catalog No. 50-197-597 Supplier Bioworld Supplier No. 201620112
Add to Cart
Add to Cart
To remove non-specific binding of proteins to magnetic bead surface and to achieve clean co-immunoprecipitation of your desired antibody or any other ligand binding. Use it with any magnetic beads experiment.Size: 2-5µm.Silica-coated magnetic beadspH stability: 4-9Supplied as 25mg/ml suspension.Requires MagneZoom paramagnetic bead station (20160000-1) for separating beads.Usage: bioWorld's products are for LABORATORY RESEARCH use only.