Learn More
Bioworld MagneZoom™ COOH, 10 mL
Catalog No. 50197591
Encompass
Carboxy-terminated Magnetic BeadsSilica-coated uniform super paramagnetic beads with surface carboxyl group. Proteins and nucleic acids can be covalently bound to these beads with the retention of biological activity.Specifications:Size: 5-10µm.COOH terminatedBinding: up to 20mg/ml.pH stability: 4-9Supplied as 25mg/ml suspension.Requires MagneZoom paramagnetic bead station (20160000-1) for bead separation.Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.
Catalog No. 50-197-591 Supplier Bioworld Supplier No. 201620082
Carboxy-terminated Magnetic BeadsSilica-coated uniform super paramagnetic beads with surface carboxyl group. Proteins and nucleic acids can be covalently bound to these beads with the retention of biological activity.Specifications:Size: 5-10µm.COOH terminatedBinding: up to 20mg/ml.pH stability: 4-9Supplied as 25mg/ml suspension.Requires MagneZoom paramagnetic bead station (20160000-1) for bead separation.Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.