Learn More
Bioworld MagneZoom™ Goat Anti-Human IgG (FC) Antibody Kit (Polyclonal IgG), 2 mL
Supplier:  Bioworld 201630161
Encompass
MagneZoom ™ Goat Anti-Human IgG (FC) Antibody Kit (Polyclonal IgG)Cat#20163016DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Human IgG (FC)Form InformationForm:Liquid SuspensionConcentration:25 mg MagneZoom™ per mL of bufferPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:2-8 °C Kit contents:• MagneZoomGoat Anti-Human IgG (Fc), 25mg/mL: 2mL• Binding/Wash Buffer • Elution Buffer
Catalog No. 50-197-620