Learn More
Bioworld MagneZoom Goat Anti-Rabbit IgG (H+L) Antibody (Polyclonal IgG), 2 mL
Catalog No. 50198356
Encompass
MagneZoom ™ Goat Anti-Rabbit IgG (H+L) Antibody (Polyclonal IgG)Cat#20330014DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Rabbit IgG (H+L)Form InformationForm:Liquid SuspensionConcentration:25 mg MagneZoom™ per mL of bufferPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:4°C
Catalog No. 50-198-356 Supplier Bioworld Supplier No. 203300142
MagneZoom ™ Goat Anti-Rabbit IgG (H+L) Antibody (Polyclonal IgG)Cat#20330014DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Rabbit IgG (H+L)Form InformationForm:Liquid SuspensionConcentration:25 mg MagneZoom™ per mL of bufferPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:4°C