Learn More
Bioworld MagneZoom™ Goat Anti-Rat IgG (FC) Antibody (Polyclonal IgG), 2 mL
Catalog No. 50198343
Encompass
MagneZoom ™ Goat Anti-Rat IgG (FC) Antibody (Polyclonal IgG)Cat#20330004DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Rat IgG (FC)Form InformationForm:Liquid SuspensionConcentration:25 mg MagneZoom™ per mL of bufferPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:4°C
Catalog No. 50-198-343 Supplier Bioworld Supplier No. 203300041
MagneZoom ™ Goat Anti-Rat IgG (FC) Antibody (Polyclonal IgG)Cat#20330004DetailsHost:GoatIsotype:IgGClass:PolyclonalImmunogen:Rat IgG (FC)Form InformationForm:Liquid SuspensionConcentration:25 mg MagneZoom™ per mL of bufferPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:4°C