Learn More
Bioworld MAGNEZOOM-PNAPEANUT AGGLU
Catalog No. 50197601
Encompass
MagneZoom-PNA (Peanut Agglutinin) MagneZoom-PNA are silica-coated super paramagnetic beads covalently linked to Peanut Agglutinin (PNA). Peanut agglutinin is a 110 kDa homotetrameric, non-glycosylated lectin with a high binding affinity ...
Catalog No. 50-197-601 Supplier Bioworld Supplier No. 201620191
MagneZoom-PNA (Peanut Agglutinin) MagneZoom-PNA are silica-coated super paramagnetic beads covalently linked to Peanut Agglutinin (PNA). Peanut agglutinin is a 110 kDa homotetrameric, non-glycosylated lectin with a high binding affinity ...