Learn More
Bioworld MagneZoom™ Protein-G Kit, 1 mL
Supplier:  Bioworld 201630111
Encompass
MagneZoom Protein G (superparamagnetic beads) of average 5 μm in diameter, covalently coated with recombinant Protein G. Protein G, a bacterial cell wall protein isolated from group G Streptococci, binds to mammalian IgGs mainly through Fc regions. Native Protein G has three IgG binding domains and also the sites for albumin and cell-surface binding. Albumin and cell-surface binding domains have been eliminated from recombinant Protein G to reduce nonspecific binding. Protein G has greater affinity than Protein A for most mammalian IgGs, especially for certain subclasses including human IgG3, mouse IgG1 and rat IgG2a. Unlike Protein A, Protein G does not bind to human IgM, IgD and IgA.Kit Components:• Protein-G MagneZoom™ (25mg/ml), 1ml• Binding/Wash Buffer • Elution Buffer • Neutralization Buffer• 1×SDS Sample Buffer This kit requires MagneZoom paramagnetic bead station (20160000-1) separating beads.
Catalog No. 50-197-615