Learn More
Bioworld Triticum vulgaris (Wheat) Lectin (WGA) - MagneZoomâ„¢
Supplier:  Bioworld 201620291
Encompass
Wheat germ -Triticum vulgaris- agglutinin (WGA) linked to MagneZoom (magnetic beads). For magnetic separation by binding specifically to cell particles, glycoconjugates and polysachharides containing N-acetylglucosamine residues.Technical Specifications:Ligand Source:Triticum vulgarisMatrix: MagneZOOM™ (Paramagnetic Silica Coated Beads)Particle size range: 2-5µmMatrix attachment: AmideBinding Capacity:1-2 mg/ mlInhibiting/Eluting Sugar: Chitin hydrolysateor GlcNAc with acidor saltCarbohydrate for Elution:.2M Chitin hydrolysateor GlcNAc with acidor saltBlood Group Specificity:A, B, OForm:25 mg/mL , PBS (1x) pH 7.4 w/ 0.01% ThimerosalpH stability: 4 – 9Storage: 2 – 8°C, DO NOT FREEZE
Catalog No. 50-197-612