Learn More
Braintree Scientific Magnification Loupe, flip up, sports frame glasses, 4x mag,
SDP
Catalog No. 501955109
Encompass
Magnifying Surgical Loupes - How to Choose the Right Product? Magnification in 2X, 3.5X, and 4 X Strengths - A 3.5x and 4X magnification surgical loupe reveals incredible detail but comes with a small field of view (30MM.) Your working distance has to be maintained more accurately, too. Choose a 4X loupe if you want to focus on a narrow work area and need to see the details that a higher magnification affords. A lower 2.5X magnification results in wider field of view (50MM) and lets you monitor a broader work area. Because of their greater depth of field, 2.5X surgical loupes allow more freedom of movement as they are less sensitive to changes in your working distance. Working Distance - Working distance is the approximate distance between your eyes and the object you work on. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5109 Supplier Braintree Scientific Supplier No. ML631
Magnifying Surgical Loupes - How to Choose the Right Product? Magnification in 2X, 3.5X, and 4 X Strengths - A 3.5x and 4X magnification surgical loupe reveals incredible detail but comes with a small field of view (30MM.) Your working distance has to be maintained more accurately, too. Choose a 4X loupe if you want to focus on a narrow work area and need to see the details that a higher magnification affords. A lower 2.5X magnification results in wider field of view (50MM) and lets you monitor a broader work area. Because of their greater depth of field, 2.5X surgical loupes allow more freedom of movement as they are less sensitive to changes in your working distance. Working Distance - Working distance is the approximate distance between your eyes and the object you work on. **Sold for research purposes only and not for human use**