Learn More
Electron Microscopy Sciences Magnifier 8x for 35 mm Film
SDP
Catalog No. 5027021
Encompass_Preferred
The 8X magnifier uses a 24 mm x 37 mm angular objective lens and a two-element achromatic lens for the eyepiece. This arrangement enables the 8X magnifier to cover the entire 24 mm x 36 mm picture area of a single frame of 35 mm film, and it is suitable for checking negatives. Engraved on the bottom edge of the transparent acrylic skirt is a 30 mm long, 1 mm division scale, which does not interfere with the field of view.
Catalog No. 50-270-21 Supplier Electron Microscopy Sciences Supplier No. 68125
The 8X magnifier uses a 24 mm x 37 mm angular objective lens and a two-element achromatic lens for the eyepiece. This arrangement enables the 8X magnifier to cover the entire 24 mm x 36 mm picture area of a single frame of 35 mm film, and it is suitable for checking negatives. Engraved on the bottom edge of the transparent acrylic skirt is a 30 mm long, 1 mm division scale, which does not interfere with the field of view.