Learn More
Glass Expansion Magnifier Inspection Tool - 10x magnification loupe with LED illumination (5190-9614). Used to inspect condition of cone orifice.
SDP
Catalog No. 502703026
Encompass_Preferred
Magnifier Inspection Tool - 10x Magnifier Inspection Tool with LED Illumination. Helps to quickly identify nebulizer or capillary tubing clogs, salt deposition at tip of nebulizer effecting aerosol generation, and to inspect ICP-MS cones for orifice damage or occlusion.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3026 Supplier Glass Expansion Supplier No. 708031923
Magnifier Inspection Tool - 10x Magnifier Inspection Tool with LED Illumination. Helps to quickly identify nebulizer or capillary tubing clogs, salt deposition at tip of nebulizer effecting aerosol generation, and to inspect ICP-MS cones for orifice damage or occlusion.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001