Learn More
Electron Microscopy Sciences EMS Magnifying Glass, Mounted to Microtome
SDP
Catalog No. 501926604
Encompass
A magnifying glass for general global slices offering approximately 1.5x magnification. Integral cold light powered directly via the Model 51-mz vibrating. For use with 5100mz or 5100mz plus.
Catalog No. 50-192-6604 Supplier Electron Microscopy Sciences Supplier No. 510013
A magnifying glass for general global slices offering approximately 1.5x magnification. Integral cold light powered directly via the Model 51-mz vibrating. For use with 5100mz or 5100mz plus.