Learn More
Electron Microscopy Sciences Green-Lite™ Light with Round LED, 6", 2.5x, Screw Base
SDP
Catalog No. 501930114
Encompass
Equipped with next generation, surface mount LEDs, these innovative illuminated magnifiers will quickly pays for themseves through energy savings.
Made in the USA they feature the highest lumen output in a lighted magnifier and reduced glare, heat, and energy consumption. User controlled LED groups shine desired level of light exactly where required, and superb dimming software via convenient, reliable membrane switch.
Features precision ground, optical grade glass and largest lens in the industry. Low voltage (15-24 v) power suplies are auto-switchable with worldwide approvals. Energy Star compliant (each unit uses 8 watts maximum). Auto shut off at 4 hour intervals to prevent energy waste while users are away. High quality die cast aluminum shades and all steel arm components for ESD safety. Virtually "maintenance zero".
Type: Screw Down Base
Catalog No. 50-193-0114 Supplier Electron Microscopy Sciences Supplier No. 6819412A
Equipped with next generation, surface mount LEDs, these innovative illuminated magnifiers will quickly pays for themseves through energy savings.
Made in the USA they feature the highest lumen output in a lighted magnifier and reduced glare, heat, and energy consumption. User controlled LED groups shine desired level of light exactly where required, and superb dimming software via convenient, reliable membrane switch.
Features precision ground, optical grade glass and largest lens in the industry. Low voltage (15-24 v) power suplies are auto-switchable with worldwide approvals. Energy Star compliant (each unit uses 8 watts maximum). Auto shut off at 4 hour intervals to prevent energy waste while users are away. High quality die cast aluminum shades and all steel arm components for ESD safety. Virtually "maintenance zero".
Type: Screw Down Base