Learn More
Electron Microscopy Sciences Green-Lite™ Light with Rectangular LED, 7"x5 ¼", 2x, Table Clamp
SDP
Catalog No. 501902121
Encompass
Equipped with next generation, surface mount LEDs, these innovative illuminated magnifiers will quickly pays for themseves through energy savings.
Made in the USA they feature the highest lumen output in a lighted magnifier and reduced glare, heat, and energy consumption. User controlled LED groups shine desired level of light exactly where required, and superb dimming software via convenient, reliable membrane switch.
Features precision ground, optical grade glass and largest lens in the industry. Low voltage (15-24 v) power suplies are auto-switchable with worldwide approvals. Energy Star compliant (each unit uses 8 watts maximum). Auto shut off at 4 hour intervals to prevent energy waste while users are away. High quality die cast aluminum shades and all steel arm components for ESD safety. Virtually "maintenance zero".
Type: Table Edge Clamp
Catalog No. 50-190-2121 Supplier Electron Microscopy Sciences Supplier No. 6819415
Equipped with next generation, surface mount LEDs, these innovative illuminated magnifiers will quickly pays for themseves through energy savings.
Made in the USA they feature the highest lumen output in a lighted magnifier and reduced glare, heat, and energy consumption. User controlled LED groups shine desired level of light exactly where required, and superb dimming software via convenient, reliable membrane switch.
Features precision ground, optical grade glass and largest lens in the industry. Low voltage (15-24 v) power suplies are auto-switchable with worldwide approvals. Energy Star compliant (each unit uses 8 watts maximum). Auto shut off at 4 hour intervals to prevent energy waste while users are away. High quality die cast aluminum shades and all steel arm components for ESD safety. Virtually "maintenance zero".
Type: Table Edge Clamp