Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Magnifying Glass, Mounted to Machine
SDP
Supplier:  Electron Microscopy Sciences 700013
Encompass_Preferred
Optional magnifying glass for EMS7000 Vibrating Microtomes has approx 2x magnification. 305mm Focal Length.
Catalog No. 50-949-754