Learn More
Cole-Parmer Kimberly-Clark OL30-06C1.5I Reader Safety Glasses, Clear Lens, Black Frame; +1.5 Diopter
Catalog No. NC9573703 Shop All Cole-Parmer Products
Encompass_Preferred
Solves the problem of workers removing safety glasses to put on reading glasses throughout the workday. Precise placement of lens insert, like a bifocal, reduces headaches and eyestrain. Compliance: ANSI Z87.1-2003
This item is not returnable. View return policy
Catalog No. NC9573703 Supplier Cole-Parmer Supplier No. 8647420
Solves the problem of workers removing safety glasses to put on reading glasses throughout the workday. Precise placement of lens insert, like a bifocal, reduces headaches and eyestrain. Compliance: ANSI Z87.1-2003