Learn More
Fine Science Tools Magnifying Loupe with Micrometer and LED
SDP
Catalog No. NC1588528
Encompass
Magnifying Loupe with Micrometer and LED. Magnification: 10xWorking Distance: 10mm (without scale). Field of View: 30mm
Catalog No. NC1588528 Supplier Fine Science Tools Supplier No. 2902610
Magnifying Loupe with Micrometer and LED. Magnification: 10xWorking Distance: 10mm (without scale). Field of View: 30mm