Learn More
MSC PRO-SAFE Magnifying Safety Glasses, 1.50 Diopter, Clear, Anti-Fog, ANSI Z87.1-2003
Catalog No. 502835387
Encompass
These magnifying safety glasses feature a 1.50 diopter clear lens with an anti-fog coating, designed for enhanced visibility in various work environments. They are constructed with a half-framed, wrap-around design and provide UV protection, meeting ANSI Z87.1-2003 compliance standards for reliable eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Clear diopter lenses provide magnification.
  • Half-framed, wrap-around design.
  • Universal size.
  • Polycarbonate lens material.
  • UV protection.
  • Meets ANSI Z87.1-2003 standards.
Catalog No. 50-283-5387 Supplier MSC Supplier No. 02395176
These magnifying safety glasses feature a 1.50 diopter clear lens with an anti-fog coating, designed for enhanced visibility in various work environments. They are constructed with a half-framed, wrap-around design and provide UV protection, meeting ANSI Z87.1-2003 compliance standards for reliable eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions.
  • Clear diopter lenses provide magnification.
  • Half-framed, wrap-around design.
  • Universal size.
  • Polycarbonate lens material.
  • UV protection.
  • Meets ANSI Z87.1-2003 standards.