Learn More
MSC MCR Safety Magnifying Safety Glasses, 2.50 Diopter, Clear Lens, Scratch-Resistant, ANSI Z87.1+
Catalog No. 502835429
Encompass
These magnifying safety glasses feature a 2.50 diopter lens, clear lenses, and scratch-resistant properties, meeting ANSI Z87.1+ standards. They offer a frameless, wrap-around design with clear frames and bayonet temples. The polycarbonate lens material provides UV protection and includes integral side shields for an unobstructed view, along with non-slip soft temple material.
  • Single-lens design with integral side shields for unobstructed view
  • Non-slip soft temple material
  • Scratch-resistant coating
  • Polycarbonate lens filters 99.9 percent of harmful UV rays
Catalog No. 50-283-5429 Supplier MSC Supplier No. 02484962
These magnifying safety glasses feature a 2.50 diopter lens, clear lenses, and scratch-resistant properties, meeting ANSI Z87.1+ standards. They offer a frameless, wrap-around design with clear frames and bayonet temples. The polycarbonate lens material provides UV protection and includes integral side shields for an unobstructed view, along with non-slip soft temple material.
  • Single-lens design with integral side shields for unobstructed view
  • Non-slip soft temple material
  • Scratch-resistant coating
  • Polycarbonate lens filters 99.9 percent of harmful UV rays