Learn More
MSC PRO-SAFE Magnifying Safety Glasses, 2.00 Diopter, Clear, Anti-Fog, ANSI Z87.1-2003
Catalog No. 502835431
Encompass
PRO-SAFE Magnifying Safety Glasses are designed for enhanced vision and protection in various work environments. These safety glasses feature a 2.00 diopter lens with an anti-fog coating for clear visibility, clear polycarbonate lenses, and a wrap-around, half-framed design. They meet ANSI Z87.1-2003 standards for eye protection.
  • 2.00 diopter lens for magnification
  • Anti-fog lens coating
  • Clear polycarbonate lenses
  • Half-framed, wrap-around design
  • Universal size
  • UV protection
  • Meets ANSI Z87.1-2003 standards
Catalog No. 50-283-5431 Supplier MSC Supplier No. 02395184
PRO-SAFE Magnifying Safety Glasses are designed for enhanced vision and protection in various work environments. These safety glasses feature a 2.00 diopter lens with an anti-fog coating for clear visibility, clear polycarbonate lenses, and a wrap-around, half-framed design. They meet ANSI Z87.1-2003 standards for eye protection.
  • 2.00 diopter lens for magnification
  • Anti-fog lens coating
  • Clear polycarbonate lenses
  • Half-framed, wrap-around design
  • Universal size
  • UV protection
  • Meets ANSI Z87.1-2003 standards